Learn More
Heidolph™ Coated Receiving Flask
Product Code. 10351442 Shop All Heidolph Instruments Products
Change view
Click to view available options
Volume (Metric):
100 mL
250 mL
500 mL
1000 mL
3000 mL
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10351442 100 mL Each
10147701 250 mL Each
10438781 500 mL Each
10381532 1000 mL Each
10779562 3000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10351442 Supplier Heidolph™ Supplier No. 5148100002
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
For use with rotary evaporators
  • With S 35/20
  • With transparent plastic coating
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material Glass
Type Receiving Flask
For Use With (Equipment) Rotary evaporators
Material (Coating) Transparent plastic
Standard Taper Joint Number Center 35/20
Volume (Metric) 100 mL