missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt Chloride Hexahydrate, MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
250 g
Unit Size:
250g
Specifications
Specifications
| CAS | 7791-13-1 |
| Quantity | 250 g |
| Molecular Formula | Cl2CoH12O6 |
| MDL Number | MFCD00149652 |
| InChI Key | GFHNAMRJFCEERV-UHFFFAOYSA-L |
| SMILES | O.O.O.O.O.O.[Cl-].[Cl-].[Co++] |
| IUPAC Name | λ²-cobalt(2+) hexahydrate dichloride |
| Molecular Weight (g/mol) | 237.92 |
| Formula Weight | 237.9 |
| Chemical Name or Material | Cobalt Chloride Hexahydrate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction