missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt(II) Carbonate Hydrate, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Cobalt(II) Carbonate Hydrate |
| CAS | 57454-67-8 |
| Molecular Formula | CCoO3 . (H2O)x |
| Quantity | 1 g |
| Synonym | Carbonic Acid Cobalt(2+) Salt (1:1) Hydrate |
| InChI Formula | InChI=1S/CH2O3.2Co.H2O/c2-1(3)4;;;/h(H2,2,3,4);;;1H2/q;;+1;/p-3 |
| SMILES | [O-]C([O-])=O.[Co][Co+].[H2O] |
| IUPAC Name | Cobalt(II) Carbonate Hydrate |
| Molecular Weight (g/mol) | 136.96 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering