Learn More
behr Labor-Technik™ COD 24 Sample Reactor
Product Code. 10421231
Change view
Click to view available options
Type: COD 24 Sample Reactor
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10421231 Type: COD 24 Sample Reactor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Product Code. 10421231 Supplier behr Labor-Technik™ Supplier No. B00217830
  • ONLINE EXCLUSIVE PRICE
    5890.50€
    3528.41€ / Pack of 1
    Save 2362.09€
    40% Off
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
R\u00E9cupel-bijdrage uitgesloten
Use these reactors for a variety of COD digestion applications. behr Labor-Technik™ COD 24 Sample Reactors feature a sophisticated design that ensures precise and uniform temperature values for all sample positions.
  • Precision heating block thermostat
  • For 24 reaction vessels
  • Heating panel for particularly even temperature distribution over all sample positions
  • Enclosure made of powder-coated steel and stainless steel.
  • The elaborate heating block insulation ensures that housing temperatures stay within safe temperature ranges.
  • Continuous grooves at the bottom of the boreholes simplify sample handling and increase user safety
  • The grooves allow for pressure equalization when the sample vessels are inserted
  • Even precisely fitting vessels can be smoothly inserted in the boreholes for user-friendly handling of the sample rack inserts
  • Remaining fluid on the outside of the reaction vessel instantly evaporates when the samples are inserted in the heated block
  • The grooves serve to discharge the vapor, thus preventing the breakage of glass due to glass vessel rupture
  • In case of vessel overflow, the sample will drain off through the grooves, reducing the risk of acid damaging the heating block
Specifications
Description Precision heating block thermostat for 24 reaction vessels RG, 20°C to 300°C
For Use With (Equipment) TRS 300 Programmable Temperature and Time Controller
Type COD 24 Sample Reactor