missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Codesoft Label Design Software

Product Code. 17164284
Change view
Click to view available options
Codesoft Label Design Software; Format: Software; For Use With (Equipment): Brady™ Label Printers; For Use With (Application): Asset Tracking, Barcode Labeling, Inventory and Inspection Labeling
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17164284 Codesoft Label Design Software; Format: Software; For Use With (Equipment): Brady™ Label Printers; For Use With (Application): Asset Tracking, Barcode Labeling, Inventory and Inspection Labeling
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17164284 Supplier Brady™ Supplier No. 196527
Add to basket
Add to basket
This item is not returnable. View return policy

Codesoft Lite allows you to create professional labels for any project, including barcoding.

Specifications

For Use With (Equipment) Brady Label Printers
Format Software
For Use With (Application) Asset Tracking, Barcode Labeling, Inventory and Inspection Labeling
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.