missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COL1A2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92790-0.02ml
This item is not returnable.
View return policy
Description
COL1A2 Polyclonal antibody specifically detects COL1A2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| COL1A2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:25-1:200, Immunohistochemistry-Paraffin | |
| alpha 2(I)-collagen, Alpha-2 type I collagen, collagen alpha-2(I) chain, collagen I, alpha-2 polypeptide, collagen of skin, tendon and bone, alpha-2 chain, collagen, type I, alpha 2, OI4, osteogenesis imperfecta type IV, type I procollagen | |
| A synthetic peptide corresponding to a sequence within amino acids 501-600 of human COL1A2 (NP_000080.2). TGDPGKNGDKGHAGLAGARGAPGPDGNNGAQGPPGPQGVQGGKGEQGPPGPPGFQGLPGPSGPAGEVGKPGERGLHGEFGLPGPAGPRGERGPPGESGAA | |
| 0.02 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction, Stem Cells | |
| 1278 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction