Learn More
Fryka™ Cold Box
Product Code. 10630366
Change view
Click to view available options
Temperature Range:
-10°C to -50°C
-50°C to -85°C
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range unitSize
10630366 -10°C to -50°C Each
10097101 -50°C to -85°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 10630366 Supplier Fryka™ Supplier No. 050B3550
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Reliable, compact, and quiet cold box designed as a desktop appliance. Fryka™ Cold Box facilitate decentralized storage of chilled items.
  • Cost-effective
  • Environmentally friendly
  • Reliable
  • Compact
  • Quiet
  • Available in different capacities and temperature ranges
Specifications
Type Cold Box
Capacity (Metric) 35 L
Temperature Range -10°C to -50°C
Dimensions (D x W x H) Interior 300 x 425 x 280 mm
Depth (Metric) Exterior 765 mm
Depth (Metric) Interior 300 mm
Height (Metric) Exterior 540 mm
Height (Metric) Interior 280 mm
Width (Metric) Exterior 580 mm
Width (Metric) Interior 425 mm
Refrigeration System Voltage 230V
Voltage 230 V
Electrical Requirements 230 V
Frequency 50 Hz
Weight (Metric) 65 kg