missing translation for 'onlineSavingsMsg'
Learn More

Colistin Sulfate (Mixture of A and B compound), TRC

Product Code. 18071900
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18071900 1 g 1g
18021830 5 g 5g
18041310 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18071900 Supplier Toronto Research Chemicals Supplier No. TRCC6415001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1264-72-8
Molecular Formula C55H100N16O20S2
Molecular Weight (g/mol) 1155.4
Synonym Colistin sulfate (7CI), Belcomycin, Colimycin sulfate, Colomycin, Colomycin syrup, Nipoxyme, Polymyxin E sulfate, Polymyxin E, sulfate (salt)
IUPAC Name Colistin sulfate
SMILES OS(O)(=O)=O.[H][C@]([C@H](O)C)(C(N[C@H](C(N[C@H]1CCNC([C@@](NC([C@@H](NC([C@@H](NC([C@@H](NC([C@H](NC([C@@H](NC1=O)CCN)=O)CC(C)C)=O)CC(C)C)=O)CCN)=O)CCN)=O)([C@H](O)C)[H])=O)=O)CCN)=O)NC([C@@H](NC(CCCCC(CC)C)=O)CCN)=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C53H100N16O13.H2O4S/c1-9-30(6)12-10-11-13-41(72)60-33(14-20-54)48(77)69-43(32(8)71)53(82)65-36(17-23-57)45(74)64-38-19-25-59-52(81)42(31(7)70)68-49(78)37(18-24-58)62-44(73)34(15-21-55)63-50(79)39(26-28(2)3)67-51(80)40(27-29(4)5)66-46(75)35(16-22-56)61-47(38)76;1-5(2,3)4/h28-40,42-43,70-71H,9-27,54-58H2,1-8H3,(H,59,81)(H,60,72)(H,61,76)(H,62,73)(H,63,79)(H,64,74)(H,65,82)(H,66,75)(H,67,80)(H,68,78)(H,69,77);(H2,1,2,3,4)/t30?,31-,32-,33+,34+,35+,36+,37+,38+,39+,40-,42+,43+;/m1./s1
Formula Weight 1266.733
Percent Purity >95%
Chemical Name or Material Colistin sulfate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.