missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen I Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-43312
This item is not returnable.
View return policy
Description
Collagen I Polyclonal antibody specifically detects Collagen I in Rat samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Collagen I | |
| Polyclonal | |
| Unconjugated | |
| PBS, pH 7.4, 50% glycerol | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 0.5 mg/mL | |
| Western Blot 0.5-2ug/mL, Immunohistochemistry 5-20 μg/mL, Immunocytochemistry/ Immunofluorescence 5-20 μg/mL, Immunoprecipitation | |
| Alpha-1 type I collagen, alpha1(I) procollagen, CAFYD, COL1A1, Collagen 1, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen alpha-1(I) chain preproprotein, collagen of skin, tendon and bone, alpha-1 chain, collagen, type I, alpha 1, EDSARTH1, EDSC, OI1, OI2, OI3, OI4, pro-alpha-1 collagen type 1, type I proalpha 1, Type I Procollagen Alpha 1 Chain | |
| Immunized with Rat Collagen I (Uniprot#: P02454 & P02466; Val1109∽Lys1372) | |
| 100 μL | |
| Cell Biology, Cellular Markers, Extracellular Matrix, Signal Transduction, Stem Cells | |
| 1277 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction