missing translation for 'onlineSavingsMsg'
Learn More

Collagen III alpha 1/COL3A1 Antibody [Biotin], Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB600-594B

Product Code. 30294396

  • 681.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Collagen III alpha 1/COL3A1 Polyclonal antibody specifically detects Collagen III alpha 1/COL3A1 in Bovine,Feline,Human,Mouse,Rat,Sheep samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Simple Western,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Collagen III alpha 1/COL3A1
Polyclonal
PBS
Rabbit
Immunogen affinity purified
RUO
Primary
Bovine, Feline, Human, Mouse, Rat, Sheep
Purified
Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Simple Western, Immunocytochemistry/Immunofluorescence
Biotin
alpha1 (III) collagen, alpha-1 type III collagen, COL3A1, Collagen 3, collagen alpha-1(III) chain, collagen, fetal, collagen, type III, alpha 1, Collagen-3, Collagen-III, collagenIII, EDS4A, EDSVASC, Ehlers-Danlos syndrome type IV, autosomal dominant, FLJ34534, PMGEDSV
Collagen III alpha 1/COL3A1 from human and bovine placenta (Uniprot: P02461)
0.1 mL
Cancer, Cell Biology, Extracellular Matrix
1281
Store at 4⪚C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.