missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen IV alpha 1 Antibody (577227) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB63082-SP
This item is not returnable.
View return policy
Description
Collagen IV alpha 1 Monoclonal specifically detects Collagen IV alpha 1 in Human samples. It is validated for Immunocytochemistry.
Specifications
| Collagen IV alpha 1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Immunocytochemistry | |
| alpha-1 chain, arresten, COL4A1 NC1 domain, collagen alpha-1(IV) chain, collagen IV, alpha-1 polypeptide, collagen, type IV, alpha 1, Collagen-4, EC 3.4.23, EC 6.3.1.2 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 577227 | |
| Unconjugated | |
| PBS with No Preservative | |
| COL4A1 | |
| Partial recombinant human Collagen IV alpha 1 protein (amino acids 1441-1669) [UniProt P02462]. | |
| 25 μg | |
| Cytoskeleton Markers, Extracellular Matrix, Stem Cell Markers | |
| 1282 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction