missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen IV Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-6586PCP
This item is not returnable.
View return policy
Description
Collagen IV Polyclonal antibody specifically detects Collagen IV in Human, Mouse, Rat, Bovine, Feline samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry Free-Floating, Gene Knock-Out, Super-Resolution Microscopy
Specifications
| Collagen IV | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bovine, Feline | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| PerCP | |
| BSVD;BSVD1;COL4A1s;Collagen alpha-1(IV) chain;PADMAL;RATOR | |
| Collagen IV from human and bovine placenta (Uniprot: P02462) | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Extracellular Matrix, Stem Cell Markers | |
| 1282 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction