Learn More
IKKγ Mouse, Unlabeled, Clone: C73-764, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 559675
Description
The transcription factor NF-κB, (nuclear factor kappa-B) is controlled by interaction with an inhibitory subunit, IκB, which restricts NF-κB to the cytoplasm. Following stimulation by various cytokines or other stimuli, IκB becomes degraded and NF-κB is released to the nucleus. The release of IκB from NF-κB is a critical step in the activation of NF-κB signal pathways. A group of proteins form an NF-κB regulatory complex, or signalsome. Two members of this complex are a pair of closely related serine/threonine kinases, IKKα and IKKβ (aka IKK-1 and IKK-2), which phosphorylate critical residues of IκB, thus targeting it for subsequent degradation. The IKK complex contains similar amounts of IKKα, IKKβ, as well as two other polypeptides, which are differentially processed forms of a third subunit, IKKγ. IKKα and IKKβ become activated following phosphorylation by upstream kinases, including NF-κB-inducing kinase (NIK) and MEKK1. IKKγ interacts preferentially with IKKβ and is required for the activation of the IKK complex. Thus the IKK kinases play an important role in the activation of NF-κB. IKKγ migrates as a doublet between 50-55kDa in SDS-PAGE.
Immunoprecipitation, Western Blotting
Specifications
| IKKγ | |
| Monoclonal | |
| 0.5mg/mL | |
| NEMO | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Western Blot | |
| C73-764 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Recombinant Human IKKγ | |
| 0.1 mg | |
| Immunology | |
| Human, Mouse | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.