missing translation for 'onlineSavingsMsg'
Learn More

Colterol Hydrochloride, TRC

Product Code. 30135240
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135240 50 mg 50mg
30133499 25 mg 25mg
17971605 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135240 Supplier Toronto Research Chemicals Supplier No. TRCC21585050MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52872-37-4
Molecular Formula C12H19NO3 • HCl
Molecular Weight (g/mol) 261.75
Synonym 4-[2-[(1,1-Dimethylethyl)amino]-1-hydroxyethyl]-1,2-benzenediol Hydrochloride, (+/-)-?-[(tert-Butylamino)methyl]-3,4-dihydroxy-benzyl Alcohol Hydrochloride, (+/-)-N-t-Butylnoradrenaline Hydrochloride, (+/-)-tert-Butyl Noradrenaline Hydrochloride, dl-1-(3,4-Dihydroxyphenyl)-2-tert-butylaminoethanol Hydrochloride, dl-N-tert-Butylnorepinephrine Hydrochloride;
IUPAC Name 4-(2-(tert-butylamino)-1-hydroxyethyl)benzene-1,2-diol hydrogen chloride
SMILES OC1=C(O)C=CC(C(CNC(C)(C)C)O)=C1.Cl

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H19NO3.ClH/c1-12(2,3)13-7-11(16)8-4-5-9(14)10(15)6-8;/h4-6,11,13-16H,7H2,1-3H3;1H
Formula Weight 261.12
Percent Purity >95%
Chemical Name or Material Colterol Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.