missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Combination pH Electrodes

Product Code. 10546381
Change view
Click to view available options
Connector Types:
BNC
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Unit Size
10546381 BNC Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10546381 Supplier Hanna Instruments™ Supplier No. HI1083B
  • ONLINE EXCLUSIVE PRICE
    1155.00€
    629.13€ / Pack of 1
    Save 525.87€
    45% Off
Estimated Shipment: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Connector Types BNC
pH Range 0 to 13
Type Combination pH Electrode
For Use With (Application) For use with biotechnology and small samples
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.