Learn More
Fisherbrand™ Combs for Horizontal Tanks MultiSUB Midi Gel Electrophoresis Unit, 2mm Thick
Product Code. 11893333 Shop All Fisherbrand Products
Change view
Click to view available options
No. of Wells:
1 Prep, 1 Marker
10
12
16
2 Prep, 2 Marker
20
25
4 Prep, 2 Marker
8
Volume (Metric) Sample:
20 μL
27 μL
32 μL
45 μL
54 μL
81 μL
153 μL
720 μL
315 μL x 8
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Wells Volume (Metric) Sample unitSize
11893333 4 Prep, 2 Marker 153 μL Each
11823313 10 54 μL Each
11853313 1 Prep, 1 Marker 720 μL Each
11893313 12 45 μL Each
11833323 16 32 μL Each
11883323 20 27 μL Each
11813333 2 Prep, 2 Marker 315 μL x 8 Each
11853333 25 20 μL Each
11813353 8 81 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 11893333
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
For use in preparatory electrophoresis and high-throughput screening.
  • Recommended to scale-up nucleic acid volumes for preparatory techniques
  • Maximise sample volume
  • Height adjustable to give user full control over well depth and sample-loading volume
  • Rigid back which prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
  • 10-sample and 20-sample combs are Multichannel pipettor compatible
Specifications
Color Blue
For Use With (Equipment) Horizontal Tanks MultiSUB Midi Gel Electrophoresis Unit
Type Comb
No. of Wells 4 Prep, 2 Marker
Quantity 1 Each
Thickness (Metric) 2 mm
Volume (Metric) Sample 153 μL