Learn More
Fisherbrand™ Combs for MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit, 1mm Thick
Artikelnummer. 11833383
Skift visning
Klik for at se tilgængelige muligheder
No. of Wells:
1 Prep, 1 Marker
10
12
14
16
18
2 Prep, 2 Marker
20
30
35
4 Prep, 2 Marker
Volume (Metric) Sample:
10 μL
11 μL
13 μL
21 μL
27 μL
29 μL
41 μL
45 μL
122 μL
225 μL
495 μL
Pakningsstørrelse:
Each
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. No. of Wells Volume (Metric) Sample Pakningsstørrelse
11833383 16 27 μL Each
11843363 10 45 μL Each
11883363 10 29 μL Each
11813373 1 Prep, 1 Marker 495 μL Each
11853373 12 41 μL Each
11893373 14 29 μL Each
11873383 18 11 μL Each
11823393 20 21 μL Each
11853393 2 Prep, 2 Marker 225 μL Each
11833403 30 13 μL Each
11873403 35 10 μL Each
11813413 4 Prep, 2 Marker 122 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Artikelnummer. 11833383
Estimated Shipment: 30-11-2026
Læg i indkøbskurv
Læg i indkøbskurv
missing translation for 'validMainframeMsg'
For use in preparatory electrophoresis and high-throughput screening.
  • Supplied as standard
  • Recommended for high resolution gels and publication quality data
  • Height adjustable to give user full control over well depth and sample-loading volume
  • Rigid back which prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
  • 10, 14, 16, 18, 28 and 30-sample combs are Multichannel pipettor compatible
Specifikationer
For Use With (Equipment) MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit
Type Comb
No. of Wells 16
Quantity 1 Each
Thickness (Metric) 1 mm
Volume (Metric) Sample 27 μL
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.