missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COMP Monoclonal Antibody (C10), Invitrogen™

Mouse Monoclonal Antibody
185.00€ - 415.00€
Specifications
| Antigen | COMP |
|---|---|
| Clone | C10 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17909872
![]() |
Invitrogen™
MA550899 |
200 μL |
415.00€
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17998922
|
Invitrogen™
MA550900 |
20 μL |
185.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
COMP is a noncollagenous extracellular matrix protein. It consists of five identical glycoprotein subunits, each with EGF like and calcium binding (thrombospondin like) domains. Oligomerization results from formation of a five stranded coiled coil and disulfides. Binding to other ECM proteins such as collagen appears to depend on divalent cations. Mutations can cause the osteochondrodysplasias pseudochondroplasia (PSACH) and multiple epiphyseal dysplasia (MED).Specifications
| COMP | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| Cartilage oligomeric matrix protein; cartilage oligomeric matrix protein (pseudoachondroplasia, epiphyseal dysplasia 1, multiple); cartilage oligomeric matrix protein(pseudoachondroplasia, epiphyseal dysplasia 1, multiple); Comp; EDM1; EPD1; MED; MGC131819; MGC149768; PSACH; pseudoachondroplasia (epiphyseal dysplasia 1, multiple); putative cartilage oligomeric matrix protein; THBS5; Thrombospondin5; Thrombospondin-5; TSP 5; TSP5 | |
| COMP | |
| IgG2b κ | |
| Protein A/G | |
| COMP |
| C10 | |
| Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Mouse | |
| Q9R0G6 | |
| 12845 | |
| Recombinant Mouse COMP protein, Gln554-Gln753 (Accession #Q9R0G6). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title