missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C3d Antibody (C3D/2891), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08764PCP
This item is not returnable.
View return policy
Description
Complement C3d Monoclonal antibody specifically detects Complement C3d in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Protein Array
Specifications
| Complement C3d | |
| Monoclonal | |
| PerCP | |
| AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product | |
| Recombinant fragment (around aa 1250-1450) of human Complement C3d protein (exact sequence is proprietary) (Uniprot: P01024) | |
| 0.1 mL | |
| Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience | |
| 718 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| ELISA, Immunohistochemistry (Paraffin), Protein Array | |
| C3D/2891 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction