missing translation for 'onlineSavingsMsg'
Learn More

Complement Component C5/C5a Antibody (06), Novus Biologicals™

Mouse Monoclonal Antibody

220.00€ - 326.00€

Specifications

Antigen Complement Component C5/C5a
Clone 06
Dilution Sandwich ELISA Capture 1:250-1:2000
Applications Sandwich ELISA Capture
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18682799
View Documents
Novus Biologicals
NBP3-06182-100ul
100 μg
326.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18682788
View Documents
Novus Biologicals
NBP3-06182-50ul
50 μg
220.00€
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Complement Component C5/C5a Monoclonal antibody specifically detects Complement Component C5/C5a in Human samples. It is validated for Sandwich ELISA Capture
Specifications

Specifications

Complement Component C5/C5a
Sandwich ELISA Capture 1:250-1:2000
Monoclonal
Purified
RUO
0.2 um filtered solution in PBS
727
IgG
Protein A purified
06
Sandwich ELISA Capture
Unconjugated
Mouse
Human
C5a;C5b;C5D;Complement C5;CPAMD4;ECLZB
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Complement Component C5/C5a (Accession#: NP_001726.2; Leu679-Arg751).
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.