Learn More
Thermo Scientific™ Complete Stainless Steel Rollout Drawer Set
Product Code. 16209581
Change view
Click to view available options
For Use With (Equipment):
11.5 cu. ft. Refrigerators
45.8 cu. ft. (1297L) Refrigerators and Freezers
No. of Drawers:
5
12
Unit Size:
Each
Pack of 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) No. of Drawers Unit Size
16209581 45.8 cu. ft. (1297L) Refrigerators and Freezers 12 Pack of 12
15206615 11.5 cu. ft. Refrigerators 5 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16209581 Supplier Thermo Scientific™ Supplier No. 6703
  • ONLINE EXCLUSIVE PRICE
    7835.69€
    7370.00€ / Pack of 12
    Save 465.69€
    5% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A convenient alternative to open-wire shelving
Choose stainless-steel Thermo Scientific™ Rollout Drawers as a convenient alternative to open-wire shelving. This is an option for Thermo Scientific™ high-performance refrigerators.
  • Stainless-steel drawers
  • Adjustable with full extension
Ordering Information:
Factory installed; must be specified when ordering the refrigerator
TRUSTED_SUSTAINABILITY
Specifications
Type Complete SS Drawer Set
Capacity (English) 45.8 cu. ft.
Capacity (Metric) 1297 L
Drawers Stainless Steel (Drawer)
No. of Drawers 12
For Use With (Equipment) 45.8 cu. ft. (1297L) Refrigerators and Freezers