Learn More
Lenz Laborglasintrumente Condensers acc. to Allihn, for Extractors, with Olives
Product Code. 10306171
Change view
Click to view available options
Standard Taper Joint No:
NS 29/32
NS 34/35
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Unit Size
10306171 NS 29/32 Each
10244101 NS 34/35 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10306171 Supplier Lenz Laborglasintrumente Supplier No. 05226004
  • ONLINE EXCLUSIVE PRICE
    127.00€
    78.58€ / Pack of 1
    Save 48.42€
    38% Off
Estimated Shipment: 21-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Allihn condensers for condensing vapours
  • With NS cone
  • Water connection: olives
  • Made of DURAN™ tubing
Specifications
Standard Taper Joint No NS 29/32
Material Duran Glass
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.