Learn More
Lenz Laborglasintrumente Condensers Acc. to Liebig, with GL Threads
Product Code. 11536794
Change view
Click to view available options
Standard Taper Joint No:
NS 13/23
NS 14/23
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Unit Size
11536794 NS 13/23 Each
11526794 NS 14/23 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11536794 Supplier Lenz Laborglasintrumente Supplier No. 05210411
  • ONLINE EXCLUSIVE PRICE
    87.50€
    72.80€ / Pack of 1
    Save 14.70€
    16% Off
Estimated Shipment: 04-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Liebig condensers acc. to DIN 12576 and in addition to DIN
  • For condensing vapours
  • With NS socket and NS cone
  • Water connection: GL 14 threads (with two threaded plastic olives)
  • Made of DURAN™ tubing
Specifications
Includes GL 14 Threads
Length (Metric) 400 mm
Standard Taper Joint No NS 13/23