Learn More
Consort Conductivity Electrode, Low Cond Graphite with Epoxy Body
Product Code. 11995478
Change view
Click to view available options
Connector Types:
1 BNC, 2 Banana
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Unit Size
11995478 1 BNC, 2 Banana Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11995478 Supplier Consort Supplier No. SK12T
  • ONLINE EXCLUSIVE PRICE
    232.60€
    180.00€ / Pack of 1
    Save 52.60€
    22% Off
View all promotions
Estimated Shipment: 13-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Consort Conductivity Electrode, Low Cond Graphite with Epoxy Body is special low-conductivity electrode with a built in ATC/temperature probe (Pt1000) constructed with dual graphite plates and supplied with a 1m cable, BNC plug and 2 banana plugs for ATC connection.
Consort offers a wide variety high quality electrochemical analytical sensors and electrode supplies.
  • Designed for laboratory, industrial, biotechnology and medical applications
  • For use with compatible benchtop or portable meters
  • While calibrating or measuring, solutions should be stirred gently to ensure accurate readings
  • Use fresh calibration solutions with values near the expected sample value
  • Rinse the electrode between measurements: first in distilled water and then with the next sample to be measured
  • Allow the electrodes sufficient time to stabilize while calibrating or measuring
  • A stability indicator on all Consort meters prompts the user when readings should be taken
Specifications
Cell Constant 0.1 cm-1
Connector Types 1 BNC, 2 Banana
For Use With (Application) Field use, Very low coductivities, Distilled water
For Use With (Equipment) Conductivity meter
Temperature (Metric) Range 0°C to 80°C
Type Low Conductivity Electrode
Diameter (Metric) Body 12 mm
Length (Metric) 110 mm
Length (Metric) Cable 1 m
Material Epoxy Body, Dual Graphite Plates
Includes ATC/temperature
Junction 2 Poles
Temperature Sensor Pt1000