Learn More
Conductivity Standard Solutions, Hanna Instruments™
Product Code. 11627030
Change view
Click to view available options
Quantity:
20 mL
Conductivity:
12,880 μS/cm
1413 μS/cm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Conductivity unitSize
11627030 20 mL 12,880 μS/cm Each
11647030 20 mL 1413 μS/cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11627030 Supplier Hanna Instruments Supplier No. HI70030P
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
A premium quality conductivity standard that is prepared against a NIST traceable potassium chloride solution.
  • Premium quality 12,880 μS/cm conductivity solution that is NIST traceable and contained in portable single-use sachets. 
  • Temperature chart of the actual μS/cm value at various temperatures printed on each bottle
  • Premium quality 12,880 μS/cm conductivity solution standardized with a conductivity meter that has been calibrated with potassium chloride NIST 2202 SRM (Standard Reference Material) 
  • Deionized water for analytical use in accordance with ISO 3696/BS3978
  • Ideal for users that do not calibrate frequently
  • Ideal for field use due to portability
Specifications
Conductivity 12,880 μS/cm
Type Calibration Standard
Industry Type Laboratory Use, Industrial Use
Quantity 20 mL