missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Connexin 30/GJB6 Antibody (CL4540) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61141AF532
This item is not returnable.
View return policy
Description
Connexin 30/GJB6 Monoclonal antibody specifically detects Connexin 30/GJB6 in Human samples. It is validated for Immunohistochemistry,Immunocytochemistry/ Immunofluorescence
Specifications
| Connexin 30/GJB6 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| connexin 30, connexin-30, Cx30, CX30gap junction protein, beta 6, DFNA3B, DFNB1B, ectodermal dysplasia 2, hidrotic (Clouston syndrome), ED2, EDH, gap junction beta-6 protein, gap junction protein, beta 6 (connexin 30), gap junction protein, beta 6, 30kDa, HEDDFNA3 | |
| This antibody was developed using a synthetic peptide derived from O95452, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| CL4540 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 10804 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction