Learn More
Connexin 30/GJB6 Antibody (CL4540) [mFluor Violet 610 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61141MFV610
Product Code. 30292106
  • 780.00€ / 0.10mL
Description
Connexin 30/GJB6 Monoclonal antibody specifically detects Connexin 30/GJB6 in Human samples. It is validated for Immunohistochemistry,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Connexin 30/GJB6
Monoclonal
mFluor Violet 610 SE
connexin 30, connexin-30, Cx30, CX30gap junction protein, beta 6, DFNA3B, DFNB1B, ectodermal dysplasia 2, hidrotic (Clouston syndrome), ED2, EDH, gap junction beta-6 protein, gap junction protein, beta 6 (connexin 30), gap junction protein, beta 6, 30kDa, HEDDFNA3
This antibody was developed using a synthetic peptide derived from O95452, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human
Purified
Immunohistochemistry, Immunocytochemistry/Immunofluorescence
CL4540
50mM Sodium Borate
Mouse
Protein A purified
RUO
10804
Store at 4⪚C in the dark.
IgG2a