missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Connexin 31 Polyclonal Antibody
GREENER_CHOICE

Product Code. 10242223
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10242223 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10242223 Supplier Invitrogen™ Supplier No. 365100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody reacts with the ~32 kDa mouse Connexin 31 on Western blots; the band observed at ~62 kDa is probably due to dimerization of Connexin 31. Several higher bands of unknown origin are observed in western blots of mouse skin homogenates.

Gap junctions are conduits that allow the direct cell-to-cell passage of small cytoplasmic molecules, including ions, metabolic intermediates, and second messengers, and thereby mediate intercellular metabolic and electrical communication. Gap junction channels consist of connexin protein subunits, which are encoded by a multigene family. GJBs (gap-junction proteins or connexins) play crucial functional roles associated with these channels. Defects in GJB3 have been linked to erythrokeratodermia variabilis (EKV) is an autosomal dominant genodermatosis characterized by transient figurate red patches or hyperkeratosis. Mutations in GJB2 have also been associated with genetically derived hearing impairments, including autosomal recessive nonsyndromic deafness.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin 31
Applications Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene GJB3
Gene Accession No. O75712, P28231
Gene Alias Cnx31; connexin 31; connexin-31; Cx31; Cxn-31; D4Wsu144e; DFNA2; DFNA2B; EKV; Gap junction beta-3 protein; gap junction membrane channel protein beta 3; gap junction protein beta 3; gap junction protein, beta 3; gap junction protein, beta 3, 31kDa; Gjb3; Gjb-3
Gene Symbols GJB3
Host Species Rabbit
Immunogen Synthetic peptide derived from the C-terminal region of mouse connexin 31 protein.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14620, 2707
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.