Learn More
Brady™ Consecutive Numbers & Letters, For Indoor Use
Product Code. 15573340 Shop All Brady Products
Change view
Click to view available options
Dimensions (L x W):
19 x 14 mm
38 x 22 mm
Unit Size:
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
15573340 38 x 22 mm Pack of 25
15523340 19 x 14 mm Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15573340 Supplier Brady™ Supplier No. 034310
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Labels that can be permanent or repositionable.
  • Labels can be permanent or repositionable; once applied, numbers & letters are there to stay - unless you want to change them
  • Made of B-499 nylon cloth with pressure sensitive adhesive that sticks to any clean, dry surface
Specifications
Material Nylon Cloth (B-499)
Adhesive Type Permanent Acrylic
Thickness 0.166 mm
Dimensions (L x W) 38 x 22 mm
Color Black on Yellow