Learn More
Liebisch™ Constanzo™ Corrosion Testing Unit
Product Code. 11910158
Change view
Click to view available options
Product Type:
Corrosion Testing System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
11910158 Corrosion Testing System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11910158 Supplier Liebisch™ Supplier No. 43026101
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Used for condensation water tests.
  • Energy-saving, heat-insulated compact chest-type design
  • Housing, testing chamber made of high-quality glass-fibre
  • Reinforced polyester resin on fully laminated steel frame with castors and adjustable feet
  • Double-wall energy-saving testing chamber cover
  • Easy handling due to double gas strut support and ergonomically formed handle
  • Function testing of filter by differential pressure measurement
  • Automatic refill system for deionized water to supply humidifier system for salt-spray testing
Specifications
Product Type Corrosion Testing System