Learn More
Contec™ CONSTIX SP-10N Sealed Polyester Swabs
BIOPHARMA
Product Code. 15805204 Shop All Contec Products
Change view
Click to view available options
Length (Metric):
70.1 mm
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
15805204 70.1 mm Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15805204 Supplier Contec™ Supplier No. SP10N
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Contec™ CONSTIX SP-10N Sealed Polyester Swabs consists of a nonwoven polyester fabric that is thermally bonded to a flex-tip polypropylene handle.
  • Nonwoven polyester head provides enhanced sorbency for non-aqueous solutions
  • Thermally sealed swab makes the swab compatible with most common solvents, such as alcohols and ketones
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Handle 2.3 mm
Diameter (Metric) Tip 1.7 mm
Length (Metric) Tip 10.8 mm
Length (Metric) 70.1 mm
Sterility Non-sterile
Width (Metric) Tip 3.3 mm
Handle Material Polypropylene