Learn More
Contec™ CONSTIX™ SP-6M Sealed Polyester Swabs
BIOPHARMA
Product Code. 16675554 Shop All Contec Products
Change view
Click to view available options
Length (Metric):
163.8 mm
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
16675554 163.8 mm Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16675554 Supplier Contec™ Supplier No. SP6M
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CONSTIX™ SP-6 sealed swab consists of 100% knitted microfiber polyester fabric that is thermally bonded to a flex-tip polypropylene handle.
The swab head is made of cleanroom laundered, double layer knitted microfiber polyester material. The swab head is constructed without adhesives or binders, making it compatible with most common solvents, such as alcohols and ketones. The flexible tip of the SP-6M swab provides a softer application of the swab head to surfaces. The swab is durable, absorbent and features extremely low particles and residue levels.
Features & Benefits
  • Flexible tip provides a softer application on surfaces
  • Polyester swab head provides enhanced sorbency, particle entrapment, and removal
TRUSTED_SUSTAINABILITY
Specifications
Length (English) 6.4 in.
Product Type Swab
Length (English) Head 0.67 in.
Length (Metric) 163.8 mm
Length (Metric) Head 17 mm
Material Polyester
Width (English) Head 0.25 in.
Width (Metric) Head 6.4 mm
Diameter (Metric) Tip 3.2 mm
Diameter (English) Tip 0.13 in.