Learn More
Thermo Scientific™ Consumables and Services Kits for Orion™ 2110XP Ammonia Analyzer
Product Code. 17138988
Change view
Click to view available options
Type:
Ammonia Calibration Kit
Ammonia Consumables Kit
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
17138988 Ammonia Calibration Kit Each
17128988 Ammonia Consumables Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17138988 Supplier Thermo Scientific™ Supplier No. 2110XPCAL
  • ONLINE EXCLUSIVE PRICE
    1560.00€
    1370.25€ / Pack of 1
    Save 189.75€
    12% Off
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Keep needed supplies and service items in stock to avoid lost productivity with Thermo Scientific™ Consumables and Services Kits for Orion™ 2110XP Ammonia Analyzer.
Keep needed supplies and service items in stock to avoid lost productivity with Thermo Scientific™ Consumables and Services Kits for the Thermo Scientific™ Orion™ 2110XP Ammonia Analyzer.
TRUSTED_SUSTAINABILITY
Specifications
Accuracy (mV) ±(0.5 mV + 0.1%)
Accuracy (Temperature) ±0.5 °C
Description Ammonia Calibration Kit with Carrying Case
Temperature (Metric) Operating -10°C to +120°C
Type Ammonia Calibration Kit
Display Type 2110XPCAL
Flow Rate 40 mL/min.
Includes 0.5mL pipette (204847-001), box of pipette tips (204846-001), ammonia calibration standards kit (181040), wash bottle and inlet filter
Inlet Pressure 8 to 100 psig
Inputs 85-132; 170-264 VAC
Outputs Alarm: 3; Relay: 3; Analog: 2; Selections: 0-20mA; 4-20 mA
Range (mV) ±1999.9 m
Relay Details Max Load: 250VAC; 30VDC; 5A
Resolution (mV) 0.1 mV
Resolution (Temperature) 0.1 °C
Response Time 2110XPCAL
Sample Inlet 0.25 NPTF Tube Fitting in.
Sample Temperature 5 to 45 °C
Show More Show Less
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.