missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liofilchem™ Contact Slide 2
Description
- Contact Slide 2 is a ready-to-use device with two different media coated onto a plastic support used for the microbial monitoring of surfaces and liquids even in the presence of residues of disinfectants. The selective medium allows the isolation and enumeration of yeasts and moulds . The other medium is used for enumeration of bacteria.
Specifications
Specifications
| Certifications/Compliance | ISO 18593:2004 |
| Content And Storage | 10°C to 25°C |
| Detects | Bacterial Count, Yeasts and Moulds |
| Format | Slide |
| Manufacturing Origin | Italy |
| Product Type | Contact Slide |
| Sterility | Non-sterile |
| Target Organism Class | Eukaryotic Microorganism, Fungi, Gram-positive Organisms, Fungi |
| Formulation | PCA + TTC + Neutralizing/Rose Bengal CAF Agar + Neutralizing |
| For Use With (Application) | For Bacterial Count and Yeasts and Moulds Detection |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?