missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liofilchem™ Contact Slide 3
PCA and TTC and Neutralizing / Vogel Johnson Agar, Flex Dip-slide with a selective medium for detection of pathogenic staphylococci and a non selective medium for total bacterial count.
Brand: Liofilchem™ 525302
Additional Details : Weight : 0.80000kg
Description
- Contact Slide 3 is a ready-to-use device with two different media coated onto a plastic support used for the microbial monitoring of surfaces and liquids even in the presence of residues of disinfectants. The selective medium allows the isolation and enumeration of coagulase-positive, mannitol-utilizing staphylococci. The other medium is used for enumeration of bacteria.
Specifications
Staphylococcus species | |
10°C to 25°C | |
Slide | |
Contact Slide | |
Gram-positive Organisms, Gram-positive Cocci, Gram-positive Cocci | |
For Bacterial Count and Staphylococci Detection | |
24 to 48hr. | |
ISO 18593:2004 |
ISO 18593:2004 | |
Bacterial Count, Staphylococci | |
Italy | |
Non-sterile | |
PCA, TTC, Neutralizing/Vogel Johnson Agar | |
At 36 ± 1°C for 24 to 48 hr. | |
20 Slides | |
Flex |