missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Contactin 4 Monoclonal Antibody (296015)

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561118

Product Code. 30535577

  • 459.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

The protein encoded by this gene is a member of the immunoglobulin superfamily. It is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contactin 4
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and no preservative
Q8IWV2
CNTN4
Mouse myeloma cell line NS0-derived recombinant human Contactin-4 isoform 1, Asp19-Gly1001.
100 μg
Primary
Human
Antibody
IgG2b
Western Blot
296015
Unconjugated
CNTN4
9630050B05; Axcam; axonal-associated cell adhesion molecule; Big-2; brain-derived immunoglobulin superfamily molecule; brain-derived immunoglobulin superfamily protein 2; CNTN4; CNTN4A; contactin 4; contactin-4; Kiaa3024; MGC33615; neural cell adhesion protein BIG-2; SCA16
Mouse
Protein A/G
RUO
152330
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.