Learn More
Buerkle™ Container Thread
Product Code. 31188862 Shop All Bürkle Products
Change view
Click to view available options
For Use With (Equipment):
Container Thread Bürkle Containers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment)
31188862 Container Thread Bürkle Containers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31188862 Supplier Buerkle™ Supplier No. 34400034
  • ONLINE EXCLUSIVE PRICE
    252.00€
    155.56€ / Pack of 1
    Save 96.44€
    38% Off
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
We will mount the container connector directly into the container for you. The thread connectors are possible anywhere. Please state the exact position on the container, if applicable.
  • Thread:G 3/4 Zoll
  • Version:External thread
  • Can be mounted on all Buerkle rectangular and round containers
TRUSTED_SUSTAINABILITY
Specifications
Type Tank Lid
For Use With (Equipment) Container Thread Bürkle Containers