Learn More
Eppendorf™ Control Panel
Product Code. 11679761 Shop All Eppendorf Products
Change view
Click to view available options
:
For use with: For Mastercycler
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11679761 For use with: For Mastercycler
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11679761 Supplier Eppendorf™ Supplier No. 6320000007
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Reproducibility, specificity and speed - are the requirements for PCR in any application
Reproducibility, specificity and speed - are the requirements for PCR in any application. The Mastercycler pro is unparalleled in its ability to fulfill these requirements. The proprietary vapo. protect™ technology reduces evaporation to a minimum. Concentrations in your PCR master mix remain consistent leading to stable and reproducible specificity. Non-specific binding is minimised beyond importance. The extremely high heating and cooling rates of the Mastercycler pro S give you the speed you need. Unsurpassed speed can be achieved with Eppendorf's Impulse PCR, a device driven hot-start function that increases heating rates to 8°C/s.
  • Ultimate reduction of evaporation
  • Extremely fast heating and cooling rates
  • Gradient blocks with SteadySlope technology
  • Intuitive graphic programming
  • Display to indicate cycler number in a network
  • Upgradable to real-time PCR*
  • Optional self-test of peltier elements
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) For Mastercycler
Product Type Control Panel
Quantity 1 Ea.