Learn More
Eppendorf™ Centrifuge 5427 R
GREENER_CHOICE
Product Code. 15825063 Shop All Eppendorf Products
Change view
Click to view available options
Electrical Requirements:
230 V, 50/60 Hz (EU)
Includes:
With Rotor FA-45-12-17
Model:
Centrifuge 5427 R
Centrifuge 5427 R (EU-IVD)
Unit Size:
1 set
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Electrical Requirements Includes Model unitSize
15825063 230 V, 50/60 Hz (EU) With Rotor FA-45-12-17 Centrifuge 5427 R 1 set
15168364 230 V, 50/60 Hz (EU) With Rotor FA-45-12-17 Centrifuge 5427 R (EU-IVD) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15825063 Supplier Eppendorf™ Supplier No. 5409000630
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The refrigerated Centrifuge 5427 R was designed for high-throughput research applications. Featuring an aerosol-tight 48-place rotor and equipped with the QuickLock lid for easy rotor lid opening and closing, it is the safe, convenient, and reliable option.
  • Rotor capacity: 48 × 1.5/2.0 mL, 12 × 5.0 mL; Eppendorf QuickLock™ rotor allows for fast and ergonomic lid locking
  • Max. speed: 25,001 × g (16,220 rpm)
  • High capacity 48-place rotor significantly increases productivity to save you valuable time
  • Soft touch one-finger closure for ergonomic operation
  • Temperature range from -11 °C to 40 °C
  • FastTemp function for quick pre-cooling of the centrifuge, e.g., from ∼ 23 °C to 4 °C in only 11 minutes
  • ECO shut-off can be programmed to engage after 1, 2, 4 or 8 hours of non-use for maximized energy savings and extended compressor life
  • Built-in condensation drain to eliminate water accumulation
  • Automatic rotor recognition and imbalance detection for maximized operational safety
  • TRUSTED_SUSTAINABILITY
    Specifications
    Capacity 12 x 2 mL
    Max. Speed 16,220 rpm
    Max. RCF 25,001 x g
    Height (English) Exterior 10 in.
    Height (Metric) Exterior 25.4 cm
    Noise Level <46 dB(A) with Rotor F-45-48-11
    Timer 10 sec. to 9:59 h
    Width (English) Exterior 12.6 in.
    Width (Metric) Exterior 31.9 cm
    Certifications/Compliance UL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers
    Includes With Rotor FA-45-12-17
    Safety Features Lid Lock, steel armoured safety chamber, view port for speed validation, lid emergency release, certified aerosol-tight rotor lids
    Electrical Requirements 230 V, 50/60 Hz (EU)
    Dimensions (L x W x H) Exterior 319 x 540 x 254 mm
    Controller Type Rotary Knob
    Model Centrifuge 5427 R
    Wattage 550 W
    Rotor Included Yes
    Refrigerated Yes
    Standards EN ISO 12100, EN 50581, EN 61010-1, EN 61010-2-020, IEC 61010-1, IEC 61010-2-020, EN 61326-1, EN 55011, EN 378-1, EN 378-2, EN 50581 (refer to manual for more details)
    Frequency 50/60 Hz
    Voltage 230 V
    Display Type Large LCD
    Temperature Range (Metric) -11°C to +40°C
    Show More Show Less
    The Centrifuge 5427 R is a non-automatic centrifuge for the separation of liquid substance mixtures from the human body and is specifically intended to be used as an accessory together with an in vitro diagnostic device and to enable its use according to its intended purpose. This product complies with the requirements of the EU In Vitro Diagnostics Regulation (IVDR) 2017/746. Eppendorf centrifuges are exclusively intended for indoor use by trained specialists.