Learn More
Corning™ Extension Collar for CoolBox™ 2XT System
Description
For use with CoolBox XT and CoolBox 2XT to extend the height when using tall tube modules. The collar is magnetized and easily adheres to the unit base.
Specifications
Specifications
| Description | Extension Collar |
| Type | Magnetized Extension Collar |
| Color | Purple |
| No. per Pack | 1/Pk. |
| No. per Case | 1/Cs. |
| For Use With (Application) | To extend height of CoolBox™ ice-free cooling systems |
| For Use With (Equipment) | CoolBox 2XT System |
| Dimensions (L x W x H) | 21.0 x 14.3 x 7.0 cm |
| Dimensions (L x W x H) Exterior | 26.5 x 20.0 x 7.0 cm |