missing translation for 'onlineSavingsMsg'
Learn More

Coomassie Blue G250, for Electrophoresis, (Page Blue G90), C.I. 42655, Fisher Chemical

Product Code. 10101520 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10101520 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10101520 Supplier Fisher Chemical Supplier No. C/P541/46

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 6104-58-1
Color Dark Blue
pH Range 5.5 to 6.5
Form Powder Solid
Melting Temperature 100°C/212°F
Molecular Weight (g/mol) 854.02g/mol
Quantity 25 g
Product Type Protein Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.