missing translation for 'onlineSavingsMsg'
Learn More

Copper Beryllium rod, alloy C17200, 2.38mm (0.094 in.) dia., Thermo Scientific™

Product Code. 11348998
Change view
Click to view available options
Quantity:
25 cm
50 cm
Unit Size:
25cm
50cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348998 25 cm 25cm
15416485 50 cm 50cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11348998 Supplier Thermo Scientific Alfa Aesar Supplier No. 042479.BU

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Copper Beryllium rod, Alloy C17200
Name Note 2.38mm (0.094 in.) dia.
Avg. Mol. Wt. or Mol. Wt. Range Cu:Be; 98.1:1.9 wt%
Quantity 25 cm
Odor Odorless
Weight ≈9.3g/25cm
Form Rod
MDL Number MFCD01320550
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.