Learn More
Copper Beryllium rod, alloy C17200, 6.35mm (0.25 in.) dia., Thermo Scientific™
Product Code. 15446485
Change view
Click to view available options
Quantity:
25 cm
Unit Size:
25cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15446485 25 cm 25cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15446485 Supplier Thermo Scientific Alfa Aesar Supplier No. 042482.BU
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Copper Beryllium rod, Alloy C17200
Name Note 6.35mm (0.25 in.) dia.
Avg. Mol. Wt. or Mol. Wt. Range Cu:Be; 98.1:1.9 wt%
Quantity 25 cm
Odor Odorless
Weight ≈66.3g/25cm
Form Rod
MDL Number MFCD01320550