missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) Bromide, 98%, Honeywell™ Fluka™

Product Code. p-7120338 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass Bottle
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 15631950

Brand: Honeywell-Fluka 21286550G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7787-70-4
Molecular Formula BrCu
Molecular Weight (g/mol) 143.45
MDL Number MFCD00010969
InChI Key NKNDPYCGAZPOFS-UHFFFAOYSA-M
Synonym copper i bromide, copper bromide cubr, copper 1+ bromide, copper monobromide, cuprousbromide, hsdb 270, cubr, copper 1 bromide
PubChem CID 24593
IUPAC Name λ¹-copper(1+) bromide
SMILES [Cu+].[Br-]

Specifications

Boiling Point 1345°C
Melting Point 492°C
Color Blue Green
Physical Form Solid
Quantity 50 g
Linear Formula CuBr
UN Number NONH for all modes of transport
Formula Weight 143.45g/mol
Percent Purity 98%
Packaging Glass Bottle
Density 4.71g/mL at 25°C
Chemical Name or Material Copper(I) Bromide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.