missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) cyanide

Product Code. p-4824696
Click to view available options
Quantity:
250 g
1 kg
2 x 2.5 kg
Unit Size:
1kg
250g
5kg
This item is not returnable. View return policy

Product Code. 11396498

Brand: Thermo Scientific Alfa Aesar 012135.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Electroplating Cu or Fe, Polymerization Catalyst | CAS: 544-92-3 | CCuN | 89.564 g/mol

Copper(I) cyanide is commonly used as a polymerization catalyst. The compound is useful in organic synthesis, as a catalyst, reagent in the preparation of nitriles(sandmeyer reaction). It is also used in electroplating copper and iron, silver plating, brass plating, and copper-tin alloy plating. It is used to make phthalocyanine dyes and pigments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-92-3
Molecular Formula CCuN
Molecular Weight (g/mol) 89.564
MDL Number MFCD00010975
InChI Key DOBRDRYODQBAMW-UHFFFAOYSA-N
Synonym copper i cyanide, cupricin, copper +1 cyanide, copper cyanide cu cn, cyanocopper, rcra waste number p029, cucn, rcra waste no. p029, cyano copper
PubChem CID 11009
IUPAC Name copper(1+);cyanide
SMILES [C-]#N.[Cu+]

Specifications

Melting Point 473°C
Color White
Physical Form Powder
Quantity 250 g
Beilstein 3587244
Solubility Information Practically insoluble in water and alcohol. Soluble in ammonium hydroxide,aqueous ammonia,pyridine and N-methylpyrrolidone.
Formula Weight 89.56
Odor Characteristic
Density 2.92
Chemical Name or Material Copper(I) cyanide
compliance-icons compliance-icons
Product Identifier
  • Copper (I) cyanide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
Supplemental information
  • MIXTURE LIST-Contains : Copper cyanide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.