missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) Oxide, Extra Pure, SLR, Fisher Chemical

Product Code. 10695292 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10695292 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10695292 Supplier Fisher Chemical Supplier No. C/8840/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Boiling Point 1800°C
Melting Point 1232°C
CAS 1317-39-1
Color Brown-Red
Physical Form Powder/Solid
Quantity 250 g
Molecular Formula Cu2H2O
MDL Number 10974
InChI Key FDARDWGARJUOKS-UHFFFAOYSA-N
SMILES [OH2--].[Cu+].[Cu+]
IUPAC Name bis(λ1-copper(1+)) oxidandiuide
Molecular Weight (g/mol) 145.11
Formula Weight 143.09g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Copper(I) oxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H318-Causes serious eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.