missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) ethylacetoacetate, 98%, BLD Pharm™

Product Code. 30780964
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30780964 25 g 25g
30778917 1 g 1g
30770288 100 g 100g
30775460 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30780964 Supplier BLD Pharm Supplier No. BD12224425g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 14284-06-1
Molecular Formula C12H18CuO6
MDL Number MFCD00017759
Quantity 25 g
SMILES CCOC1=O[Cu+2]2(O=C(C)[CH-]1)O=C(C)[CH-]C(OCC)=O2
Molecular Weight (g/mol) 321.82
Percent Purity 98%
Chemical Name or Material Copper(II) ethylacetoacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.