missing translation for 'onlineSavingsMsg'
Learn More

Copper(Ii) Oxide, ≥99.0%, ACS Reagent, Honeywell™ Fluka™

Product Code. 15642530 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15642530 500 g Poly Bottle 500g
15632530 100 g Poly Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15642530 Supplier Honeywell-Fluka Supplier No. 241741500G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1317-38-0
Molecular Formula CuO
Molecular Weight (g/mol) 79.55
MDL Number MFCD00010979
InChI Key KKCXRELNMOYFLS-UHFFFAOYSA-N
Synonym cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
PubChem CID 14829
IUPAC Name copper(2+) oxidandiide
SMILES [O--].[Cu++]

Specifications

Melting Point 1336°C
Color Black
Quantity 500 g
UN Number UN3077
Formula Weight 79.55g/mol
Percent Purity ≥99.0%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Powder
Chemical Name or Material Copper(Ii) Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.