missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Stearate (contains ~10% inorganics), TRC

Product Code. 30356919
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30356919 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30356919 Supplier Toronto Research Chemicals Supplier No. TRCC685333100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Copper(II) Stearate
CAS 660-60-6
Molecular Formula C36H70CuO4
Quantity 100 g
Synonym Cu(octanoate)2, Cupric distearate, Cupric stearate, Cupric octadecanoate, Copper(2+) stearate, Copper(II) octadecanoate
InChI Formula InChI=1S/2C18H36O2.Cu/c2*1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h2*2-17H2,1H3,(H,19,20);/q;;+2/p-2
SMILES [Cu+2].CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-]
IUPAC Name copper;octadecanoate
Molecular Weight (g/mol) 630.48
Formula Weight 629.457
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.