missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Sulfate Solution, 6.24%, Blue, Chem-Lab

Product Code. 11937433
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11937433 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11937433 Supplier Chem-Lab Supplier No. CL07.1102.0100
  • ONLINE EXCLUSIVE PRICE
    125.00€
    58.90€ / 100mL
    Save 66.10€
    52% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used as a fungicide. Copper(II) Sulfate Solution, 6.24%, Blue, Chem-Lab is a common salt of copper. Its applications include use as a fungicide, algaecide, pesticide, laboratory analytical reagent, feed supplement, zinc etching and as a mordant. According to Eur. Ph. 4(2.2.2).

Copper(II) sulfate is an inorganic compound. The pentahydrate form is the most commonly encountered salt and is bright blue. It exothermically dissolves in water. It is a common laboratory reagent.

Specifications

CAS 7758-99-8
Color Blue
Quantity 100 mL
Density 1.01g/mL
Molecular Formula CuH10O9S
MDL Number MFCD00149681
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
Solubility Information Soluble in water
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
IUPAC Name copper;sulfate;pentahydrate
Molecular Weight (g/mol) 249.68
PubChem CID 24463
ChEBI CHEBI:31440
Concentration or Composition (by Analyte or Components) 6.24%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium dichromate 0.167 mol/l
Signal Word
  • Danger||Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H340-May cause genetic defects.
  • H302-Harmful if swallowed.
  • H350-May cause cancer.
  • H315-Causes skin irritation.
  • H360-May damage fertility or the unborn child.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: potassium dichromate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.